Learn More
Biomatik Corporation ABLIM1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07692
Encompass_Preferred
The ABLIM1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Actin-binding LIM protein 1 protein (174-319AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Actin-binding LIM protein 1abLIM-1, Actin-binding LIM protein family member 1, Actin-binding double zinc finger protein, LIMAB1, Limatin, ABLIM1, ABLIM, KIAA0059, LIMAB1. UniProt: O14639. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7277