Learn More
Biomatik Corporation SLC30A7 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier: Biomatik Corporation CAC07740
Slc30A7 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC30A7. The Slc30A7 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Zinc Transporter 7 Protein (162-236Aa). Applications: ELISA, WB, IHC. Target Synonyms: SLC30A7. Accession Number: Q8NEW0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.