Learn More
Biomatik Corporation BDH1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08259
Encompass_Preferred
The BDH1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human D-beta-hydroxybutyrate dehydrogenase, mitochondrial protein (47-343AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: 3-hydroxybutyrate dehydrogenase, BDH1, BDH. UniProt: Q02338. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7843