Learn More
Biomatik Corporation C19orf47 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08268
Encompass_Preferred
The C19orf47 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Uncharacterized protein C19orf47 protein (1-281AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Uncharacterized protein C19orf47, C19orf47. UniProt: Q8N9M1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7852