Learn More
Biomatik Corporation DFNA5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08358
Encompass_Preferred
The DFNA5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Non-syndromic hearing impairment protein 5 protein (1-496AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Inversely correlated with estrogen receptor expression 1, ICERE-1, DFNA5.ICERE1. UniProt: O60443. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7941