Learn More
Biomatik Corporation F2R anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08416
Encompass_Preferred
The F2R Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Proteinase-activated receptor 1 protein (42-102AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Proteinase-activated receptor 1, PAR-1, Coagulation factor II receptor, Thrombin receptor, F2R, CF2R, PAR1, TR. UniProt: P25116. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7999