Learn More
Biomatik Corporation KAT8 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier: Biomatik Corporation CAC09207
Kat8 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KAT8. The Kat8 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Histone Acetyltransferase Kat8 Protein (1-168Aa). Applications: ELISA, WB. Target Synonyms: KAT8. Accession Number: Q9H7Z6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.