Learn More
Biomatik Corporation PLXND1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09327
Encompass_Preferred
Plxnd1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PLXND1. The Plxnd1 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Plexin-D1 Protein (1-222Aa). Applications: ELISA, IHC, IF. Target Synonyms: PLXND1. Accession Number: Q9Y4D7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-8909