Learn More
Biomatik Corporation KLHDC8A anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09472
Encompass_Preferred
Klhdc8A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to KLHDC8A. The Klhdc8A Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Kelch Domain-Containing Protein 8A Protein (1-350Aa). Applications: ELISA, WB, IHC. Target Synonyms: KLHDC8A. Accession Number: Q8IYD2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9054