Learn More
Biomatik Corporation MRGPRX2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09549
Encompass_Preferred
Mrgprx2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MRGPRX2. The Mrgprx2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Mas-Related G-Protein Coupled Receptor Member X2 Protein (286-330Aa). Applications: ELISA, WB, IF. Target Synonyms: MRGPRX2. Accession Number: Q96LB1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9131