Learn More
Biomatik Corporation C11orf68 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09713
Encompass_Preferred
The C11orf68 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human UPF0696 protein C11orf68 protein (1-251AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: UPF0696 protein C11orf68, C11orf68, BLES03, Basophilic leukemia-expressed protein Bles03, Protein p5326. UniProt: Q9H3H3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9295