Learn More
Biomatik Corporation MND1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09839
Encompass_Preferred
Mnd1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MND1. The Mnd1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Meiotic Nuclear Division Protein 1 Homolog Protein (2-205Aa). Applications: ELISA, WB, IHC. Target Synonyms: MND1. Accession Number: Q9BWT6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9421