Learn More
Biomatik Corporation FGFBP2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09904
Encompass_Preferred
The FGFBP2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Fibroblast growth factor-binding protein 2 protein (20-223AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:200-1000, IHC:1:20-1:200. Alternative Names: Fibroblast growth factor-binding protein 2, FGF-BP2, FGF-binding protein 2, FGFBP-2, 37 kDa killer-specific secretory protein, Ksp37, HBp17-related protein, HBp17-RP, FGFBP2, KSP37. UniProt: Q9BYJ0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9486