Learn More
Biomatik Corporation DSE anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09933
Encompass_Preferred
The DSE Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Dermatan-sulfate epimerase protein (23-270AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Dermatan-sulfate epimerase (EC:5.1.3.19), DS epimerase, DSE, SART2, Chondroitin-glucuronate 5-epimerase, Squamous cell carcinoma antigen recognized by T-cells 2, SART-2. UniProt: Q9UL01. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9515