Learn More
Biomatik Corporation ACSL5 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09942
Encompass_Preferred
The ACSL5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant human Long-chain-fatty-acid--CoA ligase 5 protein (33-683AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-5000, IHC:1:200-1:500. Alternative Names: Long-chain-fatty-acid--CoA ligase 5, Long-chain acyl-CoA synthetase 5, LACS 5, ACSL5, ACS5, FACL5, UNQ633/PRO1250. UniProt: Q9ULC5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9524