Learn More
Biomatik Corporation STK38L anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09962
Encompass_Preferred
Stk38L Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to STK38L. The Stk38L Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Serine; Threonine-Protein Kinase 38-Like Protein (212-464Aa). Applications: ELISA, WB, IHC. Target Synonyms: STK38L. Accession Number: Q9Y2H1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9544