Learn More
Biomatik Corporation YTHDF2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09978
Encompass_Preferred
Ythdf2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to YTHDF2. The Ythdf2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Yth Domain-Containing Family Protein 2 Protein (201-500Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: YTHDF2. Accession Number: Q9Y5A9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9560