Learn More
Biomatik Corporation CAPN6 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC10002
Encompass_Preferred
The CAPN6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Calpain-6 protein (195-494AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Calpain-6, Calpain-like protease X-linked, Calpamodulin, CalpM, CAPN6, CALPM, CANPX. UniProt: Q9Y6Q1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9584