Learn More
Biomatik Corporation Filaggrin (FLG) anti-Mouse, Rat, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21797
Encompass_Preferred
The Filaggrin (FLG) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Rat. Immunogen: FLG (Asp119-Tyr336). Applications: WB, IHC. Alternative Names: Profilaggrin. UniProt: P11088. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9770