Learn More
Biomatik Corporation Inter Alpha-Globulin Inhibitor H5 (ITIH5) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21881
Encompass_Preferred
Inter Alpha-Globulin Inhibitor H5 (Itih5) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Inter Alpha-Globulin Inhibitor H5 (ITIH5). The Inter Alpha-Globulin Inhibitor H5 (Itih5) Polyclonal Antibody has been validated for the following applications: WB. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Asp569~Gln737. Applications: WB. Target Synonyms: Inter Alpha-Globulin Inhibitor H5 (ITIH5). Accession Number: Q8BJD1. Gene ID: 209378. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9774