Learn More
Biomatik Corporation Filamin C Gamma (FLNC) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU22604
Encompass_Preferred
The Filamin C Gamma (FLNC) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: FLNC (Glu17-Lys259). Applications: WB, IHC. Alternative Names: ABP-280; ABP280A; ABPA; ABPL; FLN2; Actin Binding Protein 280; ABP-280-like protein; Actin-binding-like protein; Filamin-2. UniProt: Q14315. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9795