Learn More
Biomatik Corporation Reelin (RL) anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU23878
Encompass_Preferred
Reelin (Reln) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Reelin (RELN). The Reelin (Reln) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Glu3143~Pro3459. Applications: WB, IHC, ICC, IP. Target Synonyms: Reelin (RELN). Accession Number: Q60841. Gene ID: 19699. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9822