Learn More
Biomatik Corporation Relaxin/Insulin Like Family Peptide Receptor 1 (RXFP1) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier: Biomatik Corporation CAU25904
Relaxin/Insulin Like Family Peptide Receptor 1 (Rxfp1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Relaxin/Insulin Like Family Peptide Receptor 1 (RXFP1). The Relaxin/Insulin Like Family Peptide Receptor 1 (Rxfp1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Pro260~Met409. Applications: WB, IHC, ICC, IP. Target Synonyms: Relaxin/Insulin Like Family Peptide Receptor 1 (RXFP1). Accession Number: Q9HBX9. Gene ID: 59350. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.