Learn More
Biomatik Corporation Phospholipase A2, Lipoprotein Associated (LpPLA2) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU26917
Encompass_Preferred
Phospholipase A2 Group Vii (Lppla2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Phospholipase A2 Group VII (LpPLA2). The Phospholipase A2 Group Vii (Lppla2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Phe22~Asn441. Applications: WB, IHC, ICC, IP. Target Synonyms: Phospholipase A2 Group VII (LpPLA2). Accession Number: Q13093. Gene ID: 7941. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9939