Learn More
Biomatik Corporation Nestin (NES) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27793
Encompass_Preferred
Nestin (Nes) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Nestin (NES). The Nestin (Nes) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Arg197~Pro462. Applications: WB, IHC, ICC, IP. Target Synonyms: Nestin (NES). Accession Number: Q6P5H2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9999