Learn More
Biomatik Corporation Early Growth Response Protein 1 (EGR1) anti-Mouse, Rat, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27933
Encompass_Preferred
The Early Growth Response Protein 1 (EGR1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Rat. Immunogen: EGR1 (Gln273-Ser431). Applications: WB, IHC. Alternative Names: Zif268; AT225; G0S30; KROX24; NGFI-A; TIS8; ZIF-268; ZNF225; NGFIA; TIS8; ZENK; Nerve Growth Factor-Induced Protein A; Transcription Factor ETR103; Zinc Finger Protein 225. UniProt: P08046. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0016