Learn More
Biomatik Corporation Mucin 1 (MUC1) anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27945
Encompass_Preferred
Mucin 1 (Muc1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Mucin 1 (MUC1). The Mucin 1 (Muc1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ser1098~Leu1255. Applications: WB, IHC, ICC, IP. Target Synonyms: Mucin 1 (MUC1). Accession Number: P15941. Gene ID: 4582. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0017