Learn More
Biomatik Corporation Glutathione (GSH) anti-General, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28170
Encompass_Preferred
The Glutathione (GSH) Polyclonal Antibody is raised in Rabbit. Species Reactivity: General. Immunogen: Small Molecule, GSH conjugated to OVA. Applications: WB, IHC, ICC, IP. Alternative Names: L-Reduced Glutathione. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0034