Learn More
Biomatik Corporation Alanine Aminotransferase (ALT) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28355
Encompass_Preferred
The Alanine Aminotransferase (ALT) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: ALT (Ile255-Ser496). Applications: WB, IHC. Alternative Names: GPT; ALAT; SGPT; AAT1; GPT1; Serum Glutamic Pyruvic Transaminase; Alanine Transaminase; Glutamate pyruvate transaminase 1; Glutamic--alanine transaminase 1. UniProt: P24298. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0046