Learn More
Biomatik Corporation Hyaluronic Acid (HA) anti-General, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28389
Encompass_Preferred
Hyaluronic Acid (Ha) Polyclonal Antibody is a Universal Polyclonal Antibody to Hyaluronic Acid (HA). The Hyaluronic Acid (Ha) Polyclonal Antibody has been validated for the following applications: ELISA, CLIA / IHC-Fr, ICC, IP (predicted). Host: Rabbit. Species: Universal. Applications: ELISA, CLIA / IHC-Fr, ICC, IP (predicted). Target Synonyms: Hyaluronic Acid (HA). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0054