Learn More
Biomatik Corporation Matrix Metalloproteinase 7 (MMP7) anti-Human, Mouse, Pig, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28603
Encompass_Preferred
Matrix Metalloproteinase 7 (Mmp7) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Matrix Metalloproteinase 7 (MMP7). The Matrix Metalloproteinase 7 (Mmp7) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Lys267. Applications: WB, IHC, ICC, IP. Target Synonyms: Matrix Metalloproteinase 7 (MMP7). Accession Number: P09237. Gene ID: 4316. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0078
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.