Learn More
Biomatik Corporation Interleukin 10 (IL10) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28807
Encompass_Preferred
Interleukin 10 (Il10) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Interleukin 10 (IL10). The Interleukin 10 (Il10) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Ser19~Ser178. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 10 (IL10). Accession Number: P18893. Gene ID: 16153. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0098