Learn More
Biomatik Corporation Inhibin Beta E (INHbE) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU28856
Encompass_Preferred
Inhibin Beta E (Inhbe) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Inhibin Beta E (INHbE). The Inhibin Beta E (Inhbe) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Thr237~Ser350. Applications: WB, IHC, ICC, IP. Target Synonyms: Inhibin Beta E (INHbE). Accession Number: P58166. Gene ID: 83729. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0105