Learn More
Biomatik Corporation Angiopoietin 1 (ANGPT1) anti-Rat, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU29018
Encompass_Preferred
The Angiopoietin 1 (ANGPT1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Rat. Immunogen: ANGPT1 (Asn304-Ala450). Applications: WB, IHC. Alternative Names: ANG1; AGP1; AGPT; ANG-I. UniProt: O35460. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-151-0119