Learn More
Biomatik Corporation Platelet Derived Growth Factor AA (PDGFAA) anti-Human, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29250
Encompass_Preferred
Platelet Derived Growth Factor Aa (Pdgfaa) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Platelet Derived Growth Factor AA (PDGFAA). The Platelet Derived Growth Factor Aa (Pdgfaa) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Ser87~Thr211. Applications: WB, IHC, ICC, IP. Target Synonyms: Platelet Derived Growth Factor AA (PDGFAA). Accession Number: P04085. Gene ID: 5154. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0162