Learn More
Biomatik Corporation Matrix Metalloproteinase 9 (MMP9) anti-Human, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29261
Encompass_Preferred
Matrix Metalloproteinase 9 (Mmp9) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Matrix Metalloproteinase 9 (MMP9). The Matrix Metalloproteinase 9 (Mmp9) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Gly213~Ala399. Applications: WB. Target Synonyms: Matrix Metalloproteinase 9 (MMP9). Accession Number: P14780. Gene ID: 4318. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0163