Learn More
Biomatik Corporation Neuropilin 1 (NRP1) anti-Human, Mouse Monoclonal Antibody (100UG)
Supplier: Biomatik Corporation CAU29288
Neuropilin 1 (Nrp1) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Neuropilin 1 (NRP1). The Neuropilin 1 (Nrp1) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Cys27~Asp444. Applications: WB, IHC, ICC, IP. Target Synonyms: Neuropilin 1 (NRP1). Accession Number: O14786. Gene ID: 8829. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.