Learn More
Biomatik Corporation Myostatin (MSTN) anti-Human, Rat, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29369
Encompass_Preferred
Myostatin (Mstn) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Myostatin (MSTN). The Myostatin (Mstn) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Asp267~Ser375. Applications: WB, IHC, ICC, IP. Target Synonyms: Myostatin (MSTN). Accession Number: O14793. Gene ID: 2660. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0185