Learn More
Biomatik Corporation Matrix Metalloproteinase 2 (MMP2) anti-Human, Mouse Monoclonal Antibody (100UG)
Supplier:  Biomatik Corporation CAU29482
Encompass_Preferred
Matrix Metalloproteinase 2 (Mmp2) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Matrix Metalloproteinase 2 (MMP2). The Matrix Metalloproteinase 2 (Mmp2) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Tyr110~Cys660. Applications: WB, IHC, ICC, IP. Target Synonyms: Matrix Metalloproteinase 2 (MMP2). Accession Number: P08253. Gene ID: 4313. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0199