Learn More
Biomatik Corporation Phosphatase And Tensin Homolog (PTEN) anti-Human, Mouse, Rat, Pig, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU29991
Encompass_Preferred
Phosphatase And Tensin Homolog (Pten) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Phosphatase And Tensin Homolog (PTEN). The Phosphatase And Tensin Homolog (Pten) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Thr2~Val403. Applications: WB, IHC, ICC, IP. Target Synonyms: Phosphatase And Tensin Homolog (PTEN). Accession Number: P60484. Gene ID: 5728. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-151-0243