Learn More
Biomatik Corporation CXCL8 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier: Biomatik Corporation CAC07046
The CXCL8 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Interleukin-8 protein (23-99AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: CXCL8, IL8??IL-8??C-X-C motif chemokine 8??Emoctakin??Granulocyte chemotactic protein 1??GCP-1??Monocyte-derived neutrophil chemotactic factor??MDNCF??Monocyte-derived neutroph. UniProt: P10145. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.