Learn More
Biomatik Corporation CD46 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07202
Encompass_Preferred
The CD46 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Membrane cofactor protein (164-325AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:200-1000, IHC:1:20-1:200. Alternative Names: TLX, Trophoblast leukocyte common antigen, CD46, MCP, MIC10. UniProt: P15529. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6789