Learn More
Biomatik Corporation CTNNA1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07204
Encompass_Preferred
The CTNNA1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Catenin alpha-1 protein (1-278AA). Applications: ELISA, WB. Recommended dilution: WB: 1:500-2000. Alternative Names: Catenin alpha-1, CTNNA1, Alpha E-catenin, Cadherin-associated protein, Renal carcinoma antigen NY-REN-13. UniProt: P35221. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6791