Learn More
Biomatik Corporation C5 anti-Pig, Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07229
Encompass_Preferred
The C5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Pig, Human. Immunogen: Recombinant Pig Complement C5a anaphylatoxin protein (1-74AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: C5. UniProt: P01032. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6816
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.