Learn More
Biomatik Corporation TEAD4 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07329
Encompass_Preferred
Tead4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TEAD4. The Tead4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transcriptional Enhancer Factor Tef-3 Protein (74-434Aa). Applications: ELISA, WB, IHC. Target Synonyms: TEAD4. Accession Number: Q15561. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6915