Learn More
Biomatik Corporation Defensin Alpha 5, Paneth Cell Specific (Defa5) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU31496
Encompass_Preferred
Defensin Alpha 5, Paneth Cell Specific (Defa5) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Defensin Alpha 5, Paneth Cell Specific (DEFa5). The Defensin Alpha 5, Paneth Cell Specific (Defa5) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Asp20~Ser93. Applications: WB, IHC, ICC, IP. Target Synonyms: Defensin Alpha 5, Paneth Cell Specific (DEFa5). Accession Number: P28312. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9659