Learn More
Biomatik Corporation Ly1 Antibody Reactive Homolog (Lyar) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21090
Encompass_Preferred
Ly1 Antibody Reactive Homolog (Lyar) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Ly1 Antibody Reactive Homolog (LYAR). The Ly1 Antibody Reactive Homolog (Lyar) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Gly270~Lys388. Applications: WB, IHC, ICC, IP. Target Synonyms: Ly1 Antibody Reactive Homolog (LYAR). Accession Number: Q08288. Gene ID: 17089. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9190