Learn More
Biomatik Corporation Follistatin Like Protein 3 (Fstl3) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21809
Encompass_Preferred
Follistatin Like Protein 3 (Fstl3) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Follistatin Like Protein 3 (FSTL3). The Follistatin Like Protein 3 (Fstl3) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Val24~Val256. Applications: WB, IHC. Target Synonyms: Follistatin Like Protein 3 (FSTL3). Accession Number: Q9EQC7. Gene ID: 83554. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9220