Learn More
Biomatik Corporation Vasorin (Vasn) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU22131
Encompass_Preferred
Vasorin (Vasn) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Vasorin (VASN). The Vasorin (Vasn) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Asn298~Gly539. Applications: WB, IHC, ICC, IP. Target Synonyms: Vasorin (VASN). Accession Number: Q6EMK4. Gene ID: 114990. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9242