Learn More
Biomatik Corporation Macrophage Receptor With Collagenous Structure (Marco) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24047
Encompass_Preferred
Macrophage Receptor With Collagenous Structure (Marco) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Macrophage Receptor With Collagenous Structure (MARCO). The Macrophage Receptor With Collagenous Structure (Marco) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Gly367~Arg519. Applications: WB, IHC, ICC, IP. Target Synonyms: Macrophage Receptor With Collagenous Structure (MARCO). Accession Number: M0R9F7. Gene ID: 367391. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9331